Quality control test suspensions
 :   CROSSTEX
Quality control test suspensions
 :  CROSSTEX
Quality control test suspensions
The control suspension (growth promotion) contains <100 CFU`s/0,1 ml.
  • 2,5 ml pro vial
  • 25 dose
Product Family Options
  • Désignation
    Cdt
  • Candida albicans
    2,5 mL
    1 * 2,5 mL
    1 * 2,5 mL - SFr. 175.00
    SFr. 175.00
    1 * 2,5 mL
       SFr. 175.00
  • Pseudomonas aeruginosa
    2,5 mL
  • Staphylococcus aureus
    2,5 mL
    1 * 2,5 mL
    1 * 2,5 mL - SFr. 226.00
    SFr. 226.00
    1 * 2,5 mL
       SFr. 226.00
  • Escherichia coli
    2,5 mL
    1 * 2,5 mL
    1 * 2,5 mL - SFr. 176.00
    SFr. 176.00
    1 * 2,5 mL
       SFr. 176.00
  • Salmonella enterica
    2,5 mL
    1 * 2,5 mL
    1 * 2,5 mL - SFr. 283.00
    SFr. 283.00
    1 * 2,5 mL
       SFr. 283.00