Liquid detergent, concentrated, Contrad 70
Concentrated liquid detergent which is ideal for laboratory washers and ultrasonic cleaners. For removing soil, grit, organic contaminants, inorganic residue, chemicals, and solvents from glass, ferrous metal, ceramic and plastics.
- Phosphate- and chlorine-free
- Free-rinsing.
Frequently Bought Together