Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:beaded filament structural protein 1, filensin
- Symbole de l'antigène:BFSP1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Formulaire:liquid
- ID de gène:631
- Synonymes antigène:CP115|CTRCT33|CP94|LIFL-H
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Température de transport:4 °C
- Purification:Purified by Protein A.
- Taille:100 μg
- Cdt:100 µl
Spécifications
À propos de cet article
Filensin, also referred to as CP94, CP95, or CP97, is an eye lens fiber, membrane-associated, cytoskeletal intermediate filament (IF) protein that is required for the assembly of beaded filaments, cytoskeletal networks that are necessary for the long-term maintenance of optical clarity. Phakinin copolymerizes with Filensin to make up the filamentous structures present in the beaded filaments. Filensin is also crucial for lens development since it regulates lens fiber cell shape conformation and lens transparency. Filensin contains a C-terminal non-α-helical domain that contributes in several ways to its function. The head domain of Filensin includes a di-arginine/aromatic amino acid motif that contains a potential protein kinase A phosphorylation site.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: BFSP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat