Anti-HELLS Polyclonal Antibody Pair
1 Set
N° de catalogue ABNOH00003070-PW1
Fournisseur: Abnova
Aller à
Product Details & Documents
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-HELLS (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-HELLS (50 µg)
Type:
Antigen: HELLS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Spécifications
- Taille:1 Set
- Western blot:Yes
- ID de gène:3070
- Cat. No.:ABNOH00003070-PW1
- Clonalité:Polyclonal
- Nom de l'antigène:helicase, lymphoid-specific
- Symbole de l'antigène:HELLS
- Précipitation immunologique:Yes
- Synonymes antigène:SMARCA6|LSH|Nbla10143|PASG
- Hôte:Multiple
- Réactivité:Human
- Température de stockage:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



