Blow Lamp, X 1650
Catalog # CAMP2000026867
Anbieter: CAMPING GAZ
Technische Daten
- Désignation:Extra fine flame burner X1650
- Cdt:1
Technische Daten
Über diesen Artikel
Extra fine flame burner. Ideal for any precise work such as soldering small components, jewelry or models
- Easy screw attachment
- Gas consumption: 30 g/h