Laboratory trays
Catalog # 216-4239
Supplier: PHAGOGENE
Specifications
- Matière:PP
- l×p×h:305×255×52 mm
- Cdt:1
Specifications
About this item
Highly versatile trays, made of blue PP, ideal for a wide range of applications
- Good chemical resistance
- Good temperature resistance
- Autoclavable