Microorganisms, KWIK-STIK™
For culture purposes and quality control applications. The KWIK-STIK™ is well-known for its clever design and accurate, repeatable results. Each KWIK-STIK™ device features a single microorganism strain in a lyophilized pellet, a reservoir of hydrating fluid, and an inoculating swab. For added convenience, a peel-off identification label is included for easy documentation. The KWIK-STIK™ is available in packs of six.
- Microorganism, hydrating fluid and inoculating swab all in one self-contained device
- Does not require frozen storage conditions
- Online Certificate of Analysis
Frequently Bought Together