Anti-IRAK4 Rabbit Polyclonal Antibody
ABFRLF-PA0173
- Type d'anticorps:Primaire
- Symbole de l'antigène:IRAK4
- Clonalité:Polyclonal
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Température de stockage:Store for 1 year at –20 °C from date of shipment
- Concentration:1 mg/ml
- Immunogène:Synthetic peptide
- Purification:Protein A column
- Taille:100 µl
- Cdt:100 µl
Type: Primary
Antigen: IRAK4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat