Anti-Peroxiredoxin 6 Rabbit Polyclonal Antibody
Catalog # ABFRLF-PA0011
Supplier: AbFrontier
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Peroxiredoxin 6
- Symbole de l'antigène:PRDX6
- Clonalité:Polyclonal
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Température de stockage:Store for 1 year at –20 °C from date of shipment
- Concentration:0,5 mg/ml
- Immunogène:Recombinant human Peroxiredoxin 6 protein purified from E.coli
- Purification:Protein A column
- Taille:100 µl
- Cdt:100 µl
Specifications
About this item
Type: Primary
Antigen: Peroxiredoxin 6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat