High leverage end cutter
High leverage end cutter, chrome plated, handles insulated with multi-component grips
N° de catalogue 238-5330
Fournisseur: KNIPEX WERKE
Aller à
Product Details & Documents
Chrome vanadium heavy-duty steel, forged, oil-hardened. For heaviest duty. Chrome plated head. With two-coloured insulated multi-component grips.
- High cutting performance with minimum effort
- Suitable for soft, hard and piano wire
- Precision cutting edges additionally induction-hardened
Length: 160 mm
Spécifications
- Désignation:High leverage end cutter, chrome plated, handles insulated with multi-component grips