Anti-WT1 Mouse Monoclonal Antibody [clone: 6F-H2]
Catalog # ABNOMAB2289
Supplier: Abnova
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Wilms tumor 1
- Symbole de l'antigène:WT1
- Clonalité:Monoclonal
- Clone:6F-H2
- Conjugaison:Unconjugated
- Hôte:Mouse
- ImmunoChimie:Yes
- Précipitation immunologique:Yes
- Isotype:IgG1 kappa
- Réactivité:Human
- Western blot:Yes
- Adsorption croisée:No
- Format:Liquid
- ID de gène:7490
- Synonymes antigène:EWS-WT1|WAGR|WT33|AWT1|NPHS4|WIT-2|GUD
- Nombre d'acides aminés:1 to 181
- Storage buffer:Tris-glycine, 150 mM NaCl (0,05% sodium azide)
- Température de stockage:Store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogène:Recombinant protein corresponding to amino acids 1-181 of human WT1.
- Taille:100 μl
- Cdt:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant WT1.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:400; Western Blot: 2 ?g/ml
Type: Primary
Antigen: WT1
Clonality: Monoclonal
Clone: 6F-H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human