Anti-CD99 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-131]
Référence ABNOMAB5019
Fournisseur: Abnova
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:CD99 molecule
- Symbole de l'antigène:CD99
- Clonalité:Monoclonal
- Clone:MEM-131
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- Cytométrie en flux:Yes
- Hôte:Mouse
- Isotype:IgM
- Réactivité:Human
- Adsorption croisée:No
- Format:Liquid
- ID de gène:4267
- Synonymes antigène:MIC2Y|MIC2X|MSK5X|MIC2|HBA71
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Température de stockage:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogène:Native purified CD99 from HPB-ALL human peripheral blood leukemia T-cell line.
- Taille:100 Reactions
- Cdt:100 µg
Spécifications
À propos de cet article
Mouse monoclonal antibody raised against native CD99.
Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: CD99
Clonality: Monoclonal
Clone: MEM-131
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human