Anti-CDKN2D Mouse Monoclonal Antibody [clone: DCS-100]
Catalog # ABNOMAB2027
Supplier: Abnova
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Cyclin-dependent kinase inhibitor 2D
- Symbole de l'antigène:CDKN2D
- Clonalité:Monoclonal
- Clone:DCS-100
- Conjugaison:Unconjugated
- Hôte:Mouse
- ImmunoChimie:Yes
- Précipitation immunologique:Yes
- Isotype:IgG1
- Réactivité:Human
- Western blot:Yes
- Adsorption croisée:No
- Format:Liquid
- ID de gène:1032
- Synonymes antigène:INK4D|p19|p19-INK4D
- Storage buffer:PBS (0,08% sodium azide)
- Température de stockage:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogène:Recombinant protein corresponding to full length human CDKN2D.
- Taille:100 μg
- Cdt:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant CDKN2D.
Type: Primary
Antigen: CDKN2D
Clonality: Monoclonal
Clone: DCS-100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human