Anti-CAMK2G (phospho T286) Monoclonal Antibody [clone: 22B1]
Catalog # ABNOMAB8281
Supplier: Abnova
Specifications
- Type d'anticorps:Primaire
- Symbole de l'antigène:CAMK2G
- Clonalité:Monoclonal
- Clone:22B1
- Conjugaison:Unconjugated
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Réactivité:Rat,Mouse
- Western blot:Yes
- Adsorption croisée:No
- Format:Liquid
- Nombre d'acides aminés:281 to 294 residues surrounding T286
- Modification:phospho-Thr286
- Storage buffer:PBS (50% glycerol, 0,09% sodium azide)
- Température de stockage:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogène:Synthetic phosphopeptide corresponding to amino acids 281-294 residues surrounding T286 of CAMK2G.
- Taille:100 μg
- Cdt:100 µg
Specifications
About this item
Mouse polyclonal antibody raised against synthetic phosphopeptide of CAMK2G.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: CAMK2G (phospho T286)
Clonality: Monoclonal
Clone: 22B1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Mouse, Rat