Anti-CCDC88A Rabbit Polyclonal Antibody
Référence ABNOPAB17324
Fournisseur: Abnova
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:coiled-coil Domain Containing 88A
- Symbole de l'antigène:CCDC88A
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Réactivité:Human,Mouse
- Adsorption croisée:No
- Format:Liquid
- ID de gène:55704
- Synonymes antigène:GIRDIN|KIAA1212|GRDN|APE|GIV|HkRP1
- Storage buffer:PBS, pH 7,4 (150mM NaCl, 0,02% sodium azide, 50% glycerol)
- Température de stockage:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogène:A synthetic peptide corresponding to internal of human CCDC88A.
- Taille:100 μg
- Cdt:100 µg
Spécifications
À propos de cet article
Rabbit polyclonal antibody raised against synthetic peptide of CCDC88A.
Recommended Dilutions: Immunohistochemistry: 1:50-1:100; ELISA: 1:5000
Type: Primary
Antigen: CCDC88A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse