Anti-ZNF775 Rabbit Polyclonal Antibody
N° de catalogue ABGEAP19381B
Fournisseur: Abgent
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:zinc finger protein 775
- Symbole de l'antigène:ZNF775
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Réactivité:Human
- Western blot:Yes
- Épitope:C-terminal
- ID de gène:285971
- Nombre d'acides aminés:406 to 432
- Storage buffer:pH 7,4
- Molecular weight:59.752 kDa
- Sequence:RSPGARDTLWGRC
- Température de stockage:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.3
- Température de transport:4 °C
- Immunogène:This ZNF775 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 406-432 amino acids from the C-terminal region of human ZNF775.
- Purification:Protein A & Peptide affinity purification
- Cdt:400 µl
Spécifications
À propos de cet article
Type: Primary
Antigen: ZNF775
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human