Order Entry
Switzerland
ContactUsLinkComponent
Anti-TMPRSS6 Rabbit Polyclonal Antibody
Anti-TMPRSS6 Rabbit Polyclonal Antibody
  ABGEAP19372A
 :  Abgent
Anti-TMPRSS6 Rabbit Polyclonal Antibody
  ABGEAP19372A
 :  Abgent

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    transmembrane protease, serine 6
  • Symbole de l'antigène:
    TMPRSS6
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Épitope:
    N-terminal
  • ID de gène:
    164656
  • Synonymes antigène:
    IRIDA
  • Nombre d'acides aminés:
    85 to 114
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    90 kDa
  • Sequence:
    SQDLTRRESSAFRSEC
  • Température de stockage:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.5
  • Température de transport:
    4 °C
  • Immunogène:
    This TMPRSS6 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 85-114 amino acids from the N-terminal region of human TMPRSS6.
  • Purification:
    Protein A & Peptide affinity purification
  • Cdt:
    400 µl

 

 

Type: Primary
Antigen: TMPRSS6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human