Anti-APC Mouse Monoclonal Antibody (Unconjugated) [clone: ]
Type: Primary
Antigen: APC
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
- Catalog No:
- BLEG623402
- BLEG623401
- Symbole de l'antigène:
- APC
- APC
- Nom de l'antigène:
- Allophycocyanin
- Allophycocyanin
- Synonymes antigène:
- DP2.5|APC|FAP|FPC|GS|DP3|DP2|Adenomatous Polyposis Coli
- DP2.5|APC|FAP|FPC|GS|DP3|DP2|Adenomatous Polyposis Coli
- Type d'anticorps:
- Primaire
- Primaire
- Clonalité:
- Monoclonal
- Monoclonal
- Conjugaison:
- Unconjugated
- Unconjugated
- Réactivité:
- Mouse
- Mouse
- Hôte:
- Mouse
- Mouse
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
Frequently Bought Together