Anti-COXII subunit II Rabbit Polyclonal Antibody
Codice catalogo AGRIAS04053A
Nome del fornitore: Agrisera
Specifiche
- Type d'anticorps:Primaire
- Nom de l'antigène:COXII subunit II
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Réactivité:Plantago euryphylla,Brassica oleracea,Plantago major,A. thaliana,Silene uniflora,Silene dioica,Physcomitrella patens
- Western blot:Yes
- Adsorption croisée:No
- Température de stockage:–20 °C
- Température de transport:Ambient
- Immunogène:KLH-conugated synthetic peptide fully conserved in all available protein sequences from eudicots includingArabidopsis thaliana AtmG00160, monocots including Oryza sativa P04373 and Physcomitrella patens Q1XGA9
- Tested applications:Blue Native PAGE
- Taille:50 µg
- Cdt:50 µG
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: COXII subunit II
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Brassica oleracea, Physcomitrella patens, Plantago euryphylla, Plantago major, Silene dioica, Silene uniflora