Anti-HMGB2 Rabbit Polyclonal Antibody
Catalog # ORIGTA323053
Supplier: OriGene
Specifications
- Type d'anticorps:Primaire
- Symbole de l'antigène:HMGB2
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Adsorption croisée:No
- Formulaire:Liquid
- Nombre d'acides aminés:166 to 180
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Molecular weight:24 kDa
- Température de stockage:–20 °C
- Température de transport:–20 °C
- Immunogène:Synthetic peptide corresponding to a region derived from 166-180 amino acids of Human high mobility group box 2
- Purification:purified by antigen affinity chromatography
- Taille:100 μl
- Cdt:100 µl
Specifications
About this item
Recommended Dilutions: ELISA: 1:1000-10000; Western Blot: 1:2000-5000
Type: Primary
Antigen: HMGB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat