Heating oil cans, with sieve
HUEN821200
- Capacité:13 l
- l×p×h:505×210×400 mm
- Cdt:1
This oil can, made of HDPE, grey, could be used in industry, workshop and household.
- Have a sieve insert at the filling opening
- With screw cap at the outlet
- Resistant to oil, diesel, petrol and diluted acids