ContactUsLinkComponent
VWR Chemicals Mucap test®, Salmonella identification
VWR Chemicals Mucap test®, Salmonella identification
N° de catalogue BLIF191500
Fournisseur:  BIOLIFE ITALIANA
Numéro CAS:  
undefined
VWR Chemicals Mucap test®, Salmonella identification
N° de catalogue BLIF191500
Fournisseur:  BIOLIFE ITALIANA
Numéro CAS:  
Spécifications
  • Cdt:
    1 Vial
  • Désignation:
    Mucap test®, 8 ml for 160 tests
Spécifications
À propos de cet article
The Mucap test® allows the presumptive detection of Salmonella spp. Wood lamp with emission at 366 nm is required. Positive colonies on the plate show blue fluorescence.
  • Easy
  • Rapid