Pipette tips, MµltiFit™
Virgin PP, autoclavable.
- Positioning rings for optimal pipette fit
- 'Universal fit' with the widest range of pipette types
- Suitable for use with multi-channel pipettes
- Smooth internal surface for maximum pipetting accuracy and reproducibility even with small sample volumes
- Certified free from RNases, DNases, human DNA, PCR inhibitors and pyrogens
With a high level of volume accuracy and reliable fit, the pleasant to use and MµltiFit pipette tips represent a superb 'routine' tip for any laboratory. They are manufactured from virgin materials and guaranteed free from pyrogens.
Frequently Bought Together