Order Entry
Switzerland
ContactUsLinkComponent
Anti-IgG F(c) Goat Polyclonal Antibody (Biotin (NHS-LC))
Anti-IgG F(c) Goat Polyclonal Antibody (Biotin (NHS-LC))
Catalog # IMMRIR1928
Supplier:  ImmunoReagents
Anti-IgG F(c) Goat Polyclonal Antibody (Biotin (NHS-LC))
Catalog # IMMRIR1928
Supplier:  ImmunoReagents

Specifications

  • Type d'anticorps:
    Secondaire
  • Nom de l'antigène:
    Immunoglobulin G
  • Symbole de l'antigène:
    IgG F(c)
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Biotin (NHS-LC)
  • ELISA:
    Yes
  • Cytométrie en flux:
    Yes
  • Hôte:
    Goat
  • ImmunoChimie:
    Yes
  • ImmunoFluorescence:
    Yes
  • Précipitation immunologique:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Mouse
  • Western blot:
    Yes
  • Épitope:
    F(c)
  • Adsorption croisée:
    Yes
  • Format:
    Purified IgG
  • Formulaire:
    Clear, colourless liquid
  • Fragment d'anticorps:
    F(c)
  • Storage buffer:
    10 mM sodium phosphate, 0,15 M sodium chloride, pH 7,2, 1 % (w/v) BSA, protease/IgG free
  • Température de stockage:
    2...8 °C
  • Concentration:
    1,50 mg/ml (E 1% at 280 nm = 13,0)
  • Température de transport:
    2...8 °C
  • Immunogène:
    Mouse IgG Fc
  • Tested applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Affinity purified using solid phaseMouse IgG
  • Taille:
    2,0 mg
  • Cdt:
    1 mg

Specifications

About this item

ImmunoReagents’ Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualised, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Ordering information: Products can by customised based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse