Order Entry
Switzerland
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (DyLight® 550)
Anti-IgG Goat Polyclonal Antibody (DyLight® 550)
Catalog # IMMRIR1658
Supplier:  ImmunoReagents
undefined
Anti-IgG Goat Polyclonal Antibody (DyLight® 550)
Catalog # IMMRIR1658
Supplier:  ImmunoReagents

Specifications

  • Type d'anticorps:
    Secondaire
  • Nom de l'antigène:
    Immunoglobulin G
  • Symbole de l'antigène:
    IgG
  • Clonalité:
    Polyclonal
  • Conjugaison:
    DyLight® 550
  • ELISA:
    Yes
  • Cytométrie en flux:
    Yes
  • Hôte:
    Goat
  • ImmunoChimie:
    Yes
  • ImmunoFluorescence:
    Yes
  • Précipitation immunologique:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Adsorption croisée:
    No
  • Format:
    Purified IgG
  • Formulaire:
    Lyophilized
  • Storage buffer:
    10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
  • Température de stockage:
    2…8 °C
  • Concentration:
    1,0 mg/ml (E 1% at 280 nm = 13,0)
  • Température de transport:
    2…8 °C
  • Immunogène:
    Human IgG
  • Tested applications:
    Fluoresence ELISA, Fluorescence-Activated Cell Sorting
  • Purification:
    Affinity purified using solid phaseHuman IgG
  • Taille:
    1,0 mg
  • Cdt:
    1 mg

Specifications

About this item

The ImmunoReagents DyLight® 550 conjugates provide vibrant orange-to-red fluorescence with better performance than other rhodamine derivatives, including Alexa Fluor 555, TRITC and Cy3 for fluorescent applications. DyLight® 550 conjugates have tight excitation/emission spectra that are ideal for multicolor applications. DyLight® 550 conjugated secondary antibodies are used in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 562 nm / 576 nm Emission Color = Orange to Red (Similar Dyes: Alexa Fluor 555, Cy3, TRITC).

  • Origin: US

ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.
Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human