Specifications
- Type d'anticorps:Secondaire
- Nom de l'antigène:Immunoglobulin G
- Symbole de l'antigène:IgG
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Cytométrie en flux:Yes
- Hôte:Chicken
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Précipitation immunologique:Yes
- Isotype:IgY
- Réactivité:Rabbit
- Western blot:Yes
- Adsorption croisée:Yes
- Format:Purified IgG
- Formulaire:Clear, colorless liquid, 0,2 µm filtered
- Storage buffer:10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Température de stockage:2...8 °C
- Concentration:>4,5 mg/ml (E 1% at 280 nm = 13,2)
- Température de transport:2...8 °C
- Immunogène:Rabbit IgG
- Tested applications:Fluorescence-activated cell sorting, immunodiffusion, labelling
- Purification:Affinity purified using solid phase Rabbit IgG
- Taille:1,0 mg
- Cdt:1 mg
Specifications
About this item
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.
- Origin: US
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Rabbit