Order Entry
Switzerland
ContactUsLinkComponent
Anti-UCN Rabbit Polyclonal Antibody
Anti-UCN Rabbit Polyclonal Antibody
Catalog # USBIU2575-04D
Supplier:  US Biological
Anti-UCN Rabbit Polyclonal Antibody
Catalog # USBIU2575-04D
Supplier:  US Biological

Specifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Urocortin
  • Symbole de l'antigène:
    UCN
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Rat
  • Format:
    Serum
  • Formulaire:
    lyophilized powder
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Taille:
    50 µl
  • Cdt:
    50 µl

Specifications

About this item

Recommended dilution: Immunofluorescence: 1:200; Immunochemistry: 1:500

Type: Primary
Antigen: UCN (urocortin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat