Anti-TRPM8 Rabbit Polyclonal Antibody
USBIT8666-38C
- Type d'anticorps:Primaire
- Nom de l'antigène:transient receptor potential cation channel, subfamily M, member 8
- Symbole de l'antigène:TRPM8
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Rat
- Formulaire:lyophilized powder
- Nombre d'acides aminés:656 to 680
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified
- Taille:200 µg
- Cdt:200 µG
Type: Primary
Antigen: TRPM8 (transient receptor potential cation channel, subfamily M, member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat