Anti-STAP1 Goat Polyclonal Antibody
USBIS7954-60
- Type d'anticorps:Primaire
- Nom de l'antigène:Signal transducing adaptor family member 1
- Symbole de l'antigène:STAP1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Goat
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Formulaire:liquid
- Storage buffer:Tris-saline, 0,02% sodium azide, 0,5% BSA.
- Température de stockage:Cool
- Concentration:~0,5 mg/ml
- Température de transport:Ice
- Purification:purified
- Taille:100 µg
- Cdt:100 µG
Type: Primary
Antigen: STAP1 (signal transducing adaptor family member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human