Anti-RNF39 Goat Polyclonal Antibody
USBIR2031-93B
- Type d'anticorps:Primaire
- Nom de l'antigène:Ring Finger Protein 39
- Symbole de l'antigène:RNF39
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Goat
- Réactivité:Pig,Human,Rat
- Western blot:Yes
- Formulaire:liquid
- Storage buffer:Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Température de stockage:Cool
- Concentration:~0,5 mg/ml
- Température de transport:Ice
- Purification:purified by immunoaffinity chromatography
- Taille:100 µg
- Cdt:100 µG
Recommended dilution: ELISA: 1:16000; Western Blot: 1-3?g/ml
Type: Primary
Antigen: RNF39 (ring finger protein 39)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Pig, Rat