Order Entry
Switzerland
ContactUsLinkComponent
Anti-PRMT5 Rabbit Polyclonal Antibody
Catalog # USBIP9004-25C
Supplier:  US Biological
undefined
Anti-PRMT5 Rabbit Polyclonal Antibody
Catalog # USBIP9004-25C
Supplier:  US Biological

Specifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Protein arginine methyltransferase 5
  • Symbole de l'antigène:
    PRMT5
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Épitope:
    C-terminal
  • Formulaire:
    liquid
  • Storage buffer:
    PBS, 0,09% sodium azide.
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Purification:
    purified by ammonium sulfate precipitation
  • Taille:
    200 µl
  • Cdt:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: PRMT5 (protein arginine methyltransferase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human