Anti-IKBKB Chicken Polyclonal Antibody
Catalog # USBII3000-23
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Inhibitor of Kappa Light Polypeptide Gene Enhancer in B-cells, kinase beta
- Symbole de l'antigène:IKBKB
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Chicken
- ImmunoChimie:Yes
- Isotype:IgY
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Formulaire:liquid
- Nombre d'acides aminés:310 to 376
- Storage buffer:PBS, pH 7,2, 0,02% sodium azide.
- Température de stockage:Cool
- Concentration:~1 mg/ml
- Température de transport:Ice
- Purification:purified by immunoaffinity chromatography
- Taille:50 µg
- Cdt:50 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:500; Immunochemistry: 1:200;
Type: Primary
Antigen: IKBKB (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat