Order Entry
Switzerland
ContactUsLinkComponent
Anti-IgG, Goat Polyclonal Antibody
Anti-IgG, Goat Polyclonal Antibody
  USBII1904-53G
Anti-IgG, Goat Polyclonal Antibody
  USBII1904-53G

 

  • Type d'anticorps:
    Primaire
  • Symbole de l'antigène:
    IgG
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Goat
  • Réactivité:
    Mouse
  • Formulaire:
    liquid
  • Storage buffer:
    10mMsodium phosphate, 0,15M sodium chloride, pH 7,2, 0,05% sodium azide.
  • Température de stockage:
    Cool
  • Concentration:
    ~1 mg/ml
  • Température de transport:
    Ice
  • Taille:
    1 mg
  • Cdt:
    1 mg

 

 

Type: Primary
Antigen: IgG,
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse