Anti-Gorasp-2 Rabbit polyclonal antibody
Catalog # USBIG8567-07
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Symbole de l'antigène:GORASP2
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Zebrafish
- Western blot:Yes
- Épitope:C-terminal
- Formulaire:liquid
- Storage buffer:40mMMOPS, pH 7,5, 0,1% BSA, 0,1% sodium azide, 50% glycerol
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by immunoaffinity chromatography
- Taille:250 µl
- Cdt:250 µl
Specifications
About this item
Recommended dilution: ELISA: 1:5000-1:20000
Type: Primary
Antigen: Gorasp-2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish