Anti-CXCR4 Rabbit Polyclonal Antibody
Catalog # USBIC8350-06B
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:chemokine (C-X-C motif) receptor 4
- Symbole de l'antigène:CXCR4
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Réactivité:Rat,Mouse
- Western blot:Yes
- Formulaire:lyophilized powder
- Storage buffer:0,1% sodium azide.
- Température de stockage:Cool
- Concentration:~1 mg/ml
- Température de transport:Ice
- Purification:purified by Protein A chromatography
- Taille:200 µg
- Cdt:200 µG
Specifications
About this item
Recommended dilution: Western Blot: 1:2000
Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat