Anti-ZNF667 Rabbit Polyclonal Antibody
Catalog # USBI146459
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:zinc finger protein 667
- Symbole de l'antigène:ZNF667
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Formulaire:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Température de stockage:Cool
- Concentration:~1 mg/ml
- Température de transport:Ice
- Purification:purified by immunoaffinity chromatography
- Taille:100 µg
- Cdt:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 2.5 ?g/ml; Immunofluorescence: 20 ?g/ml
Type: Primary
Antigen: ZNF667 (zinc finger protein 667)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat