Anti-GAPDH Chicken Polyclonal Antibody
Catalog # USBI146381
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Glyceraldehyde-3-phosphate dehydrogenase
- Symbole de l'antigène:GAPDH
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Chicken
- Isotype:IgY
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Épitope:N-terminal
- Formulaire:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Température de stockage:Cool
- Concentration:~1 mg/ml
- Température de transport:Ice
- Purification:purified by immunoaffinity chromatography
- Taille:100 µg
- Cdt:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-2?g/ml
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat