Anti-TOMM70A Rabbit Polyclonal Antibody
Catalog # USBI146360
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae)
- Symbole de l'antigène:TOMM70A
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Épitope:internal
- Formulaire:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Température de stockage:Cool
- Concentration:~1 mg/ml
- Température de transport:Ice
- Purification:purified by immunoaffinity chromatography
- Taille:100 µg
- Cdt:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 1-2?g/ml; Immunofluorescence: 20 ?g/ml; Immunochemistry: 2.5 ?g/ml
Type: Primary
Antigen: TOMM70A (translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat