Anti-IL1RL1 Rabbit Polyclonal Antibody
USBI146011
- Type d'anticorps:Primaire
- Nom de l'antigène:Interleukin 1 Receptor-like 1
- Symbole de l'antigène:IL1RL1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Épitope:N-terminal
- Formulaire:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Température de stockage:Cool
- Concentration:~1 mg/ml
- Température de transport:Ice
- Purification:purified by immunoaffinity chromatography
- Taille:100 µg
- Cdt:100 µG
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 2?g/ml; Immunofluorescence: 20 ?g/ml
Type: Primary
Antigen: IL1RL1 (interleukin 1 receptor-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse