Order Entry
Switzerland
ContactUsLinkComponent
Anti-ATXN3 Rabbit Polyclonal Antibody
Anti-ATXN3 Rabbit Polyclonal Antibody
  USBI144213
undefined
Anti-ATXN3 Rabbit Polyclonal Antibody
  USBI144213

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    ataxin 3
  • Symbole de l'antigène:
    ATXN3
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Formulaire:
    lyophilized powder
  • Storage buffer:
    5mgBSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mgthimerosal, 0,05mg NaN3.
  • Température de stockage:
    Cool
  • Concentration:
    ~0,5 mg/ml (after reconstitution)
  • Température de transport:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Taille:
    100 µg
  • Cdt:
    100 µG

 

 

Recommended dilution: Immunochemistry: 0.5 - 1 ?g/ml; Western Blot: 0.1 - 0.5 ?g/ml

Type: Primary
Antigen: ATXN3 (ataxin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat