Anti-TPD52 Rabbit Polyclonal Antibody
USBI134631
- Type d'anticorps:Primaire
- Nom de l'antigène:Tumor Protein D52
- Symbole de l'antigène:TPD52
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Formulaire:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MDRGEQGLLRTDPVPEEGEDVAATISATETLSEEEQEELRRELAKVEEEIQTLSQVLAAKEKHLAEIKRKLGINSLQELKQNIAKGWQDVTATSAYKKTSETLSQAGQKASAAFSSVGSVITKKLEDVKNSPTFKSFEEKVENLKSKVGGTKPAGGDFGEVLNSAANASATTTEPLPEKTQESL
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by Protein A chromatography
- Taille:100 µg
- Cdt:100 µG
Type: Primary
Antigen: TPD52 (tumor protein D52)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human