Anti-PPP2R2C Mouse Polyclonal Antibody
Katalognr. USBI131715
Anbieter: US Biological
Technische Daten
- Type d'anticorps:Primaire
- Nom de l'antigène:protein phosphatase 2, regulatory subunit B, gamma
- Symbole de l'antigène:PPP2R2C
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Mouse
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Formulaire:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MGKRDTADIISTVEFNHTGELLATGDKGGRVVIFQREPESKNAPHSQGEYDVYSTFQSHEPEFDYLKSLEIEEKINKIKWLPQQNAAHSLLSTNDKTIKLWKITERDKRPEGYNLKDEEGKLKDLSTVTSLQVPVLKPMDLMVEVSPRRIFANGHTYHINSISVNSDCETYMSADDLRINLWHLAITDRSFNIVDIKPANMEDLTEVITASEFHPHHCNLFVYSSSKGSLRLCDMRAAALCDKHSKLFEEPEDPSNRSFFSEIISSVSDVKFSHSGRYMLTRDYLTVKVWDLNMEARPIETYQVHDYLRSKLCSLYENDCIFDKFECAWNGSDSVIMTGAYNNFFRMFDRNTKRDVTLEASRESSKPRAVLKPRRVCVGGKRRRDDISVDSLDFTKKILHTAWHPAENIIAIAATNNLYIFQDKVNSDMH
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by Protein A chromatography
- Taille:50 µg
- Cdt:50 µG
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: PPP2R2C (protein phosphatase 2, regulatory subunit B, gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human