Anti-JAG1 Rat Polyclonal Antibody
USBI044968
- Type d'anticorps:Primaire
- Nom de l'antigène:jagged 1
- Symbole de l'antigène:JAG1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rat
- Western blot:Yes
- Formulaire:liquid
- Storage buffer:0,2um-filtered PBS, pH 7,4
- Température de stockage:Cool
- Concentration:1 mg/ml
- Température de transport:Ice
- Purification:purified by Protein A chromatography
- Taille:100 µg
- Cdt:100 µG
Type: Primary
Antigen: JAG1 (jagged 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: