Anti-FLT4 Rabbit Polyclonal Antibody
Catalog # USBI043744
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Fms-related tyrosine kinase 4
- Symbole de l'antigène:FLT4
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human
- Épitope:C-terminal
- Formulaire:liquid
- Nombre d'acides aminés:1122 to 1152
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by Protein G chromatography
- Taille:200 µl
- Cdt:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-100
Type: Primary
Antigen: FLT4 (fms-related tyrosine kinase 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human