Anti-ULK3 Rabbit Polyclonal Antibody
USBI043604
- Type d'anticorps:Primaire
- Nom de l'antigène:Unc-51 like kinase 3
- Symbole de l'antigène:ULK3
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Épitope:internal
- Formulaire:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by Protein A chromatography
- Taille:200 µl
- Cdt:200 µl
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100
Type: Primary
Antigen: ULK3 (unc-51 like kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse