Anti-TSC2 Rabbit Polyclonal Antibody
Catalog # USBI043345
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:tuberous sclerosis 2
- Symbole de l'antigène:TSC2
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Mouse
- Formulaire:liquid
- Modification:phospho-Ser1155
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Température de stockage:Cool
- Température de transport:Ice
- Tested applications:Dot Blot
- Purification:purified by Protein A chromatography
- Taille:200 µl
- Cdt:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Dot blot 1:500
Type: Primary
Antigen: TSC2 (tuberous sclerosis 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse