Anti-CILP2 Rabbit Polyclonal Antibody
200 µl
N° de catalogue USBI033914
Fournisseur: US Biological
Aller à
Product Details & Documents
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Type: Primary
Antigen: CILP2 (cartilage intermediate layer protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
Spécifications
- Taille:200 µl
- Western blot:Yes
- Type d'anticorps:Primaire
- Formulaire:liquid
- Purification:purified by Protein A chromatography
- Symbole de l'antigène:CILP2
- Isotype:IgG
- Conjugaison:Unconjugated
- Réactivité:Human
- Température de stockage:Cool
- Cat. No.:USBI033914
- Clonalité:Polyclonal
- Épitope:internal
- Nombre d'acides aminés:832 to 862
- Nom de l'antigène:cartilage intermediate layer Protein 2
- Hôte:Rabbit
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Température de transport:Ice
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



