Anti-NR1H4 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNB400-153F
Supplier: Novus Biologicals
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:nuclear receptor subfamily 1, group H, member 4
- Symbole de l'antigène:NR1H4
- Clonalité:Polyclonal
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- Hôte:Rabbit
- ImmunoFluorescence:Yes
- Réactivité:Human
- Western blot:Yes
- ID de gène:9971
- Synonymes antigène:HRR-1|BAR|FXR|RIP14|HRR1
- Température de stockage:Cool
- Température de transport:Ice
- Taille:100 μL
- Cdt:0,1 mL
Specifications
About this item
Type: Primary
Antigen: NR1H4 (nuclear receptor subfamily 1, group H, member 4)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human