Anticorps polyclonal anti-PITPNC1 de souris
Référence ABCAAB68013-50
Fournisseur: Abcam
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:phosphatidylinositol transfer Protein, cytoplasmic 1
- Symbole de l'antigène:PITPNC1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Mouse
- Western blot:Yes
- Température de stockage:Cool
- Température de transport:Ice
- Taille:50 µg
- Type de conditionnement:Vial
- Cdt:50 µG
Spécifications
À propos de cet article
Mouse polyclonal to PITPNC1.
Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: PITPNC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human