Order Entry
Switzerland
ContactUsLinkComponent
Anticorps monoclonal de souris anti-CD36 [clone : FA6-152]
Anticorps monoclonal de souris anti-CD36 [clone : FA6-152]
Catalog # ABCAAB17044-50
Supplier:  Abcam
undefined
Anticorps monoclonal de souris anti-CD36 [clone : FA6-152]
Catalog # ABCAAB17044-50
Supplier:  Abcam

Specifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    CD36 molecule (thrombospondin receptor)
  • Symbole de l'antigène:
    CD36
  • Clonalité:
    Monoclonal
  • Clone:
    FA6-152
  • Conjugaison:
    Unconjugated
  • Cytométrie en flux:
    Yes
  • Hôte:
    Mouse
  • ImmunoChimie:
    Yes
  • ImmunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Seq:
    1202
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Taille:
    50 µg
  • Type de conditionnement:
    Vial
  • Cdt:
    50 µG

Specifications

About this item

Mouse monoclonal [FA6-152] to CD36.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: FA6-152
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human